missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FTSJ3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584559
This item is not returnable.
View return policy
Description
Immunogen sequence: KAVLQEEQAN LWFSKGSFAG IEDDADEALE ISQAQLLFEN RRKGRQQQQK QQLPQTPPSC LKTE Highest antigen sequence indentity to the following orthologs: Mouse - 69%, Rat - 67%.
Although the function of this gene is not known, the existence of this gene is supported by mRNA and EST data. A possible function of the encoded protein can be inferred from amino acid sequence similarity to the E.coli FtsJ protein and to a mouse protein possibly involved in embryogenesis.
Specifications
| FTSJ3 | |
| Polyclonal | |
| Unconjugated | |
| FTSJ3 | |
| 2'-O-ribose RNA methyltransferase SPB1 homolog; 2'-O-ribose RNA methyltransferase SPB1 homolog {ECO:0000255; AA537063; AU045295; C79843; D11Ertd400e; ectoplacental cone, invasive trophoblast giant cells, extraembryonic ectoderm and chorion sequence 3; Epcs3; FLJ20062; FtsJ homolog 3; FtsJ homolog 3 (E. coli); FtsJ RNA 2'-O-methyltransferase 3; FtsJ RNA methyltransferase homolog 3; FTSJ3; HAMAP-Rule:MF_03163}; pre-rRNA 2'-O-ribose RNA methyltransferase FTSJ3; pre-rRNA 2'-O-ribose RNA methyltransferase FTSJ3; pre-rRNA processing protein FTSJ3; pre-rRNA processing protein FTSJ3; pre-rRNA processing protein FTSJ3 {ECO:0000255; protein ftsJ homolog 3; Putative rRNA methyltransferase 3; putative rRNA methyltransferase 3 {ECO:0000255; rRNA (uridine-2'-O-)-methyltransferase 3; SB92; SPB1; SPB1 RNA methyltransferase homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 117246 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IY81 | |
| FTSJ3 | |
| Recombinant protein corresponding to Human FTSJ3. Recombinant protein control fragment (Product #RP-107179). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction