Learn More
FTY720 (S)-Phosphate, MedChemExpress
Product Code. 16429067
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16429067 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16429067 Supplier Medchem Express Supplier No. HY15382
MedChemExpress FTY720 (S)-Phosphate is an agonist of S1P receptor 1 (S1PR1), used in the research of acute inflammatory diseases such as acute lung injury.
FTY720 (S)-Phosphate is an agonist of S1P receptor 1 (S1PR1), used in the research of acute inflammatory diseases such as acute lung injury.
Specifications
Chemical Name or Material FTY720 (S)-Phosphate
Physical Form Powder
CAS 402616-26-6
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C19H34NO5P
For Use With (Application) COVID-19-immunoregulation
Synonym (S)-FTY720P (S)-FTY720 phosphate
Solubility Information H2O : < 0.1 mg/mL (insoluble)
SMILES OC[C@](CCC1=CC=C(CCCCCCCC)C=C1)(N)COP(O)(O)=O
Molecular Weight (g/mol) 387.45
Formula Weight 387.45
Percent Purity 98.0%
Grade Research
Show More Show Less
Research purposes only