missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUBP1 Antibody (14F5), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59448MFV450
This item is not returnable.
View return policy
Description
FUBP1 Monoclonal antibody specifically detects FUBP1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| FUBP1 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry | |
| 14F5 | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| far upstream element (FUSE) binding protein 1, far upstream element binding protein, far upstream element-binding protein 1, FBPDNA helicase V, FUBP, FUSE-binding protein 1, hDH V | |
| Recombinant human FUBP1 (279-448aa) purified from E.coli | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 8880 | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction