Learn More
Invitrogen™ FUBP1 Recombinant Rabbit Monoclonal Antibody (1G6X5)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA542907
Product Code. 17789463
  • € 524.00 / 100µL
Description
Positive test controls include: Mouse brain, Rat brain. Immunogen sequence: YQQQAQPPPA APAGAPTTTQ TNGQGDQQNP APAGQVDYTK AWEEYYKKMG QAVPAPTGAP PGGQPDYSAA WAEYYRQQAA YYAQTSPQGM PQHPPAPQGQ.
This gene encodes a ssDNA binding protein that activates the far upstream element (FUSE) of c-myc and stimulates expression of c-myc in undifferentiated cells. Regulation of FUSE by FUBP occurs through single-strand binding of FUBP to the non-coding strand. This protein has been shown to function as an ATP-dependent DNA helicase.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FUBP1
Recombinant Monoclonal
0.39 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q32PX7, Q91WJ8, Q96AE4
FUBP1
A synthetic peptide corresponding to a sequence within amino acids 545-644 of human FUBP1 (Q96AE4).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1G6X5
Unconjugated
FUBP1
9530027K12Rik; D3Ertd330e; DNA helicase V; EMBL:AAI07943.1, ECO:0000312; EMBL:AAI07943.1}; far upstream element (FUSE) binding protein 1; far upstream element (FUSE) binding protein 4; far upstream element binding protein 1; far upstream element-binding protein; far upstream element-binding protein 1; far upstream element-binding protein 1 {ECO:0000250; FBP; FBP {ECO:0000250; Fubp; Fubp1; fubp1 {ECO:0000312; Fubp4; FUSE binding protein 1; FUSE-binding protein 1; FUSE-binding protein 1 {ECO:0000250; hDH V; RGD:1591892}; UniProtKB:Q96AE4, ECO:0000312; UniProtKB:Q96AE4}
Rabbit
Affinity Chromatography
RUO
51886, 654496, 8880
-20°C, Avoid Freeze/Thaw Cycles