missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUBP3 Monoclonal Antibody (OTI9B2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF812228
This item is not returnable.
View return policy
Description
FUBP3 Monoclonal antibody specifically detects FUBP3 in Human samples. It is validated for Western BlotSpecifications
| FUBP3 | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9Y5Y7 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 10894 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI9B2 | |
| Unconjugated | |
| Lyve1 | |
| A330051M14Rik; AV006371; far upstream element (FUSE) binding protein 3; far upstream element binding protein 3; far upstream element-binding protein 3; FBP3; FUBP3; FUSE-binding protein 3; MAP2 RNA trans-acting protein 2; Marta2 | |
| Human recombinant protein fragment corresponding to amino acids 20-238 of human LYVE1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction