missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FUBP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115218
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FBP3.
May interact with single-stranded DNA from the far-upstream element (FUSE). May activate gene expression.
Specifications
| FUBP3 | |
| Polyclonal | |
| Unconjugated | |
| FUBP3 | |
| A330051M14Rik; AV006371; far upstream element (FUSE) binding protein 3; far upstream element binding protein 3; far upstream element-binding protein 3; FBP3; FUBP3; FUSE-binding protein 3; MAP2 RNA trans-acting protein 2; Marta2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 320267, 8939 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q96I24 | |
| FUBP3 | |
| A synthesized peptide derived from human FUBP3(Accession Q96I24), corresponding to amino acid residues R408-S458. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction