missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fucosyltransferase 3/FUT3 Antibody (2-25LE; same as LWB01), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14260PE
This item is not returnable.
View return policy
Description
Fucosyltransferase 3/FUT3 Monoclonal antibody specifically detects Fucosyltransferase 3/FUT3 in Human, Guinea Pig samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Fucosyltransferase 3/FUT3 | |
| Monoclonal | |
| PE | |
| CD174, FT3B, FucT-III, LE, Les | |
| Mucin isolated from a human ovarian cyst fluid (Uniprot: P21217) | |
| 0.1 mL | |
| Primary | |
| Human, Guinea Pig | |
| Purified |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 2-25LE; same as LWB01 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 2525 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction