missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fumarase Antibody (3F6), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59442F
This item is not returnable.
View return policy
Description
Fumarase Monoclonal antibody specifically detects Fumarase in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifications
| Fumarase | |
| Monoclonal | |
| FITC | |
| EC 4.2.1.2, Fumarase, fumarate hydratase, fumarate hydratase, mitochondrial, HLRCC, LRCC, MCL, MCUL1 | |
| Recombinant human Fumarase (44-510aa) purified from E. coli | |
| 0.1 mL | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 2271 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Flow Cytometry, ELISA, Immunofluorescence | |
| 3F6 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction