missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fumarase Antibody (OTI1F10), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70733PCP
This item is not returnable.
View return policy
Description
Fumarase Monoclonal specifically detects Fumarase in Human, Mouse, Rat, Canine samples. It is validated for Flow Cytometry.
Specifications
| Fumarase | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| FH | |
| Full length recombinant protein of human Fumarase (NP_000134) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG1 |
| Flow Cytometry | |
| OTI1F10 | |
| Flow Cytometry | |
| EC 4.2.1.2, Fumarase, fumarate hydratase, fumarate hydratase, mitochondrial, HLRCC, LRCC, MCL, MCUL1 | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 2271.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction