missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fumarase Monoclonal Antibody (GT548)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA527745

Product Code. 16352254

  • € 566.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive Control: 293T, mouse liver, rat liver Predicted Reactivity: Zebrafish (90%), Pig (98%), Chicken (91%), Sheep (99%), Chimpanzee (99%), Bovine (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is an enzymatic component of the tricarboxylic acid cycle, or Krebs cycle, and catalyzes the formation of L-malate from fumarate. It exists in both a cytosolic form and an N-terminal extended form, differing only in the translation start site used. The N-terminal extended form is targeted to the mitochondrion, where the removal of the extension generates the same form as in the cytoplasm. It is similar to some thermostable class II fumarases and functions as a homotetramer. Mutations in this gene can cause fumarase deficiency and lead to progressive encephalopathy.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fumarase
Monoclonal
1 mg/mL
PBS with no preservative
P07954, P14408, P97807
FH, Fh1
Recombinant protein encompassing a sequence within the center region of human Fumarate hydratase. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG2b
Western Blot, Immunocytochemistry
GT548
Unconjugated
FH
EF-3; Fh; Fh1; Fh-1; FMRD; fumarase; fumarate hydratase; fumarate hydratase 1; fumarate hydratase, mitochondrial; HLRCC; LRCC; MCL; MCUL1
Mouse
Protein G
RUO
14193, 14194, 2271, 24368
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato