missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Fumarase Monoclonal specifically detects Fumarase in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
| Antigen | Fumarase |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 3F6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1048, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Formulation | Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide |
| Gene Alias | EC 4.2.1.2, Fumarase, fumarate hydratase, fumarate hydratase, mitochondrial, HLRCC, LRCC, MCL, MCUL1 |
| Gene Symbols | FH |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?