missing translation for 'onlineSavingsMsg'
Learn More

Fumarylacetoacetate hydrolase Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-48736

 View more versions of this product

Product Code. 18675437

  • € 559.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Fumarylacetoacetate hydrolase Polyclonal antibody specifically detects Fumarylacetoacetate hydrolase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Fumarylacetoacetate hydrolase
Polyclonal
Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:500 - 1:1000
beta-diketonase, EC 3.7.1.2, FAA, fumarylacetoacetase, Fumarylacetoacetate hydrolase, fumarylacetoacetate hydrolase (fumarylacetoacetase)
This antibody was developed against a recombinant protein corresponding to amino acids: INLSVNLKGEGMSQAATICKSNFKYMYWTMLQQLTHHSVNGCNLRPGDLLASGTISGPEPENFGSMLELSWKGTKPIDLGNGQTRKFLLDGD
0.1 mL
Lipid and Metabolism
2184
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.2), 40% Glycerol
Rabbit
Immunogen affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.