missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUNDC1 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18250G
This item is not returnable.
View return policy
Description
FUNDC1 Polyclonal antibody specifically detects FUNDC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| FUNDC1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry | |
| FUN14 domain containing 1, FUN14 domain-containing protein 1, MGC51029 | |
| Synthetic peptide from the N-terminal of Human FUNDC1 | |
| 0.1 mL | |
| Cell Biology, Endocrinology | |
| 139341 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| DyLight 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction