missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FUNDC1 Recombinant Rabbit Monoclonal Antibody (8W4R3)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551458
This item is not returnable.
View return policy
Description
FUNDC1 Recombinant Monoclonal Antibody for IHC (P), ELISA
Acts as an activator of hypoxia-induced mitophagy, an important mechanism for mitochondrial quality control.
Specifications
| FUNDC1 | |
| Recombinant Monoclonal | |
| 0.29 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q5BJS4 | |
| FUNDC1 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Rat | |
| Antibody | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 8W4R3 | |
| Unconjugated | |
| FUNDC1 | |
| 1500005J14Rik; 1810033P05Rik; FUN14 domain containing 1; FUN14 domain-containing protein 1; fundc1; hypothetical protein LOC436984; si:ch211-222m21.2; zgc:92600 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 363442 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction