Learn More
Bochem™ Funnels
Click to view available options
Diameter (Metric) Stem:
17/13mm
20mm
Height (Metric):
115 mm
135 mm
Unit Size:
Each
Description
- Special funnel
- For powder
- Straight tube
- Made from 18/10 stainless steel
Specifications
Specifications
| Diameter (Metric) Stem | 20mm |
| Material | Steel |
| Height (Metric) | 135 mm |