Learn More
Furin Antibody (1C3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26563-100ul
 View more versions of this product
Product Code. 30043470
  • € 533.00 / 100µL
Description
Furin Monoclonal antibody specifically detects Furin in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Furin
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot, ELISA, Immunohistochemistry
1C3
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Dibasic-processing enzyme, EC 3.4.21, EC 3.4.21.75, FURdibasic processing enzyme, furin (paired basic amino acid cleaving enzyme), furin, membrane associated receptor protein, PACEFES upstream region, paired basic amino acid cleaving enzyme (furin, membrane associated receptorprotein), Paired basic amino acid residue-cleaving enzyme, PCSK3furin, proprotein convertase subtilisin/kexin type 3, SPC1
A synthesized peptide derived from Human Furin [UniProt P09958]
100 μL
Golgi Apparatus Markers, Stem Cell Signaling Pathway
5045
Store at -20 to -70C. Avoid freeze-thaw cycles.