Learn More
Furin Antibody (222722), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB1503V
Product Code. 30059028
  • € 854.00 / 0.50mL
Description
Furin Monoclonal antibody specifically detects Furin in Human samples. It is validated for Western Blot, Immunoprecipitation, Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Furin
Monoclonal
Alexa Fluor 405
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Human
Purified
Western Blot, Immunoprecipitation, Flow Cytometry, CyTOF
222722
Western Blot, Immunoprecipitation, Intracellular Staining by Flow Cytometry, CyTOF-ready
Dibasic-processing enzyme, EC 3.4.21, EC 3.4.21.75, FURdibasic processing enzyme, furin (paired basic amino acid cleaving enzyme), furin, membrane associated receptor protein, PACEFES upstream region, paired basic amino acid cleaving enzyme (furin, membrane associated receptorprotein), Paired basic amino acid residue-cleaving enzyme, PCSK3furin, proprotein convertase subtilisin/kexin type 3, SPC1
Mouse myeloma cell line NS0-derived recombinant human Furin, Asp108-Glu715, Accession # NP_002560
0.5 mL
Golgi Apparatus Markers, Stem Cell Signaling Pathway
5045
Store at 4C in the dark.
IgG2b