Learn More
Carbolite™ FuRNAce Bwf11/13
Brand:  Carbolite™ CWF-B 11/13
Additional Details : Weight : 70.00000kg
Product Code. 12973751
  • € 7180.00 / Each
Estimated Shipment: 28-05-2024
to see stock.
Add to basket
Description
Description
Ashing and burn off furnace requires controller 201PID or 2416CG or 2408CP (specify with order) 230V 50/60Hz 1100°C 12.9L Carbolite