Learn More
Carbolite™ Furnace with Type R Temperature Sensor
Brand:  Carbolite™ RHF1608-230SN+&02-301
Additional Details : Weight : 100.00000kg
 View more versions of this product
Product Code. 10766904
  • € 27170.00 / Each
Estimated Shipment: 08-07-2024
to see stock.
Specifications
Compliance to BSEN 61010
8 L
Furnace
270 x 170 x 170 mm
170 mm
170 mm
270 mm
230 V
50/60 Hz
301 PID Controller
1600°C
For use in quality control, heat treatment and research of ceramic materials.
505 mm
705 mm
675 mm
230 V 50/60 Hz 8000 w
8000 w