Learn More
Fusaric Acid, TRC
Product Code. 30133586
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133586 1 g 1g
30135189 250 mg 250mg
30010792 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133586 Supplier Toronto Research Chemicals Supplier No. TRCF8654001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 536-69-6
Molecular Formula C10 H13 N O2
Molecular Weight (g/mol) 179.22
Synonym Fusaric acid (6CI), Picolinic acid, 5-butyl- (7CI,8CI), 5-Butyl-2-pyridinecarboxylic acid, 5-Butyl-2-picolinic acid, 5-Butylpicolinic acid, 5-n-Butylpyridine-2-carboxylic acid, NSC 135043, NSC 19870
IUPAC Name 5-butylpyridine-2-carboxylic acid
SMILES O=C(O)C1=NC=C(C=C1)CCCC
Specifications
Quantity 1 g
InChI Formula InChI=1S/C10H13NO2/c1-2-3-4-8-5-6-9(10(12)13)11-7-8/h5-7H,2-4H2,1H3,(H,12,13)
Formula Weight 179.0946
Chemical Name or Material Fusaric Acid