missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUT10 Antibody (4H3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00084750-M04
This item is not returnable.
View return policy
Description
FUT10 Monoclonal antibody specifically detects FUT10 in Human samples. It is validated for Western Blot, ELISA
Specifications
| FUT10 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| alpha 1,3-fucosyl transferase, alpha-(1,3)-fucosyltransferase 10, EC 2.4.1, EC 2.4.1.-, Fucosyltransferase 10, fucosyltransferase 10 (alpha (1,3) fucosyltransferase), Fucosyltransferase X, Fuc-TX, FucT-X, Galactoside 3-L-fucosyltransferase 10, MGC11141 | |
| FUT10 (AAH04884, 1 a.a. ∽ 92 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MEEAPTHLNSFLKKEGLTFNRKRKWELDSYPIMLWWSPLTGETGRLGQCGADACFFTINRTYLHHHMTKAFLFYGKQDFRLSPLFAVVFLQS | |
| 0.1 mg | |
| Neuroscience | |
| 84750 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA | |
| 4H3 | |
| Western Blot 1:500, ELISA | |
| AAH04884 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction