Learn More
FUT9 Mouse, anti-Human, CoraLite Plus 488, Clone: 5D4G6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-60230-20UL
Description
The FUT9 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FUT9. This antibody recognizes human, pig, mouse, rat antigen. The FUT9 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| FUT9 | |
| 5D4G6 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Pig, Mouse, Rat | |
| 155 kDa | |
| IgG2a |
| Immunofluorescence | |
| 1000 μg/mL | |
| Alpha (1,3) fucosyltransferase, Fuc TIX, Fucosyltransferase 9, Fucosyltransferase IX, FucT IX, FUT9 | |
| 20 μL | |
| 10690 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |