Learn More
FUT9 Mouse, anti-Human, CoraLite Plus 488, Clone: 5D4G6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-60230-20UL
 View more versions of this product
Product Code. 18074255
  • € 145.00 / 20µL
Description
The FUT9 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human FUT9. This antibody recognizes human, pig, mouse, rat antigen. The FUT9 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
FUT9
5D4G6
CoraLite Plus 488
Mouse
Primary
Human, Pig, Mouse, Rat
155 kDa
IgG2a
Immunofluorescence
1000 μg/mL
Alpha (1,3) fucosyltransferase, Fuc TIX, Fucosyltransferase 9, Fucosyltransferase IX, FucT IX, FUT9
20 μL
10690
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.