missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FUT9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567133
This item is not returnable.
View return policy
Description
Immunogen sequence: ADSFIHVEDYN SPSELAKYLK EVDKNNKLYL SYFNWRKDFT VNLPRFWESH ACLACDHVKR HQEYKSVGNL EKWF Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.
Transfers a fucose to lacto-N-neotetraose but not to either alpha2,3-sialyl lacto-N-neotetraose or lacto-N-tetraose. Can catalyze the last step in the biosynthesis of Lewis antigen, the addition of a fucose to precursor polysaccharides.
Specifications
| FUT9 | |
| Polyclonal | |
| Unconjugated | |
| FUT9 | |
| AI746471; Alpha-(1,3)-fucosyltransferase 9; AU067636; fucosyltransferase 9; fucosyltransferase 9 (alpha (1,3) fucosyltransferase); fucosyltransferase IX; Fuc-TIX; fucT-IX; FUT9; Galactoside 3-L-fucosyltransferase; mFuc-TIX; mFUT9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10690 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y231 | |
| FUT9 | |
| Recombinant Human FUT9. Recombinant protein control fragment (Product #RP-107780). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction