missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FxCycle™ Violet Ready Flow™ Reagent

Product Code. 15993332
Change view
Click to view available options
Quantity:
120 assays
Unit Size:
120 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15993332 120 assays 120 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15993332 Supplier Invitrogen™ Supplier No. R37166

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

FxCycle™ Violet Ready Flow™ Reagent is a bright, easy-to-use dsDNA stain for cell cycle analysis in fixed cells using flow cytometry.

It is excitable by both UV and 405 nm lasers with an emission peak at 461 nm.

• Stable at room temperature
• Convenient, ready-to-use format—no need to dilute, weigh, or pipette
• Rapid staining of DNA in fixed cells
• No RNase treatment required

FxCycle Violet Ready Flow Reagent is a ready-to-use, direct-to-sample DNA stain. Add 2 drops from the dropper bottle directly into 1 x 106 fixed cells, incubate, and analyze.

As a cell-impermeant dye, FxCycle Violet Ready Flow Reagent enters the cell after fixation and binds double-stranded DNA specifically, eliminating the need for RNase treatment. The dye can be detected when excited with a UV or 405 nm laser, and the emission captured at a peak of 461 nm. FxCycle Violet Ready Flow Reagent is an ideal choice for DNA content analysis and can be included in multi-color cell-cycle studies.

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Content And Storage 2 dropper bottles, store at room temperature
Product Type Reagent
Dye Type Cell-Impermeant
Emission 358/461
Product Line FxCycle Ready Flow
Quantity 120 assays
Sub Cellular Localization Nucleus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.