Learn More
FXN Mouse anti-Human, Clone: OTI1C12, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF504254
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
FXN is a mitochondrial protein which belongs to the FRATAXIN family. The protein functions in regulating mitochondrial iron transport and respiration. The expansion of intronic trinucleotide repeat GAA results in Friedreich ataxia.Specifications
| FXN | |
| Monoclonal | |
| Unconjugated | |
| FXN | |
| CyaY, FA, FARR, FRDA, X25 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 2395 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C12 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q16595 | |
| FXN | |
| Full length human recombit protein of human FXN produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.