missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FXR1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22246AF532
This item is not returnable.
View return policy
Description
FXR1 Polyclonal antibody specifically detects FXR1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| FXR1 | |
| Polyclonal | |
| Western Blot | |
| fragile X mental retardation syndrome-related protein 1, fragile X mental retardation, autosomal homolog 1, FXR1P, hFXR1p | |
| The immunogen this antibody was made to, maps to a region between residue 571 and 621 of human Fragile X Mental Retardation, Autosomal Homolog 1 using the numbering given in entry NP_005078.2 (GeneID 8087). | |
| 0.1 mL | |
| Apoptosis, Immune System Diseases, Immunology | |
| 8087 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction