Learn More
FXYD3 Antibody (OTI4E3), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-71219AF750
Product Code. 18112675
  • € 751.00 / 0.10mL
Description
FXYD3 Monoclonal specifically detects FXYD3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FXYD3
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
FXYD3
Full length recombinant protein of human FXYD3 (NP_005962) produced in HEK293T cells.
Primary
Human
IgG2a
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
OTI4E3
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Chloride conductance inducer protein Mat-8, FXYD domain containing ion transport regulator 3, FXYD domain-containing ion transport regulator 3, Mammary tumor 8 kDa protein, MAT8MAT-8, Phospholemman-like, phospholemman-like protein, PLMLMGC111076
Mouse
0.1 mL
5349
Store at 4°C in the dark.