missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FXYD3 Antibody (OTI4E3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-01991
This item is not returnable.
View return policy
Description
FXYD3 Monoclonal specifically detects FXYD3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| FXYD3 | |
| Monoclonal | |
| 1.11 mg/mL | |
| Western Blot 1:500-2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:150 | |
| FXYD3 | |
| Full length human recombinant protein of human FXYD3(NP_005962) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| 5349 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI4E3 | |
| Unconjugated | |
| Chloride conductance inducer protein Mat-8, FXYD domain containing ion transport regulator 3, FXYD domain-containing ion transport regulator 3, Mammary tumor 8 kDa protein, MAT8MAT-8, Phospholemman-like, phospholemman-like protein, PLMLMGC111076 | |
| Mouse | |
| 7.1 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction