missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FXYD7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541057
This item is not returnable.
View return policy
Description
Peptide sequence: MATPTQTPTK APEEPDPFYY DYNTVQTVGM TLATILFLLG ILIVISKKVK Sequence homology: Cow: 77%; Human: 100%.
The specific function of this protein remains unknown.This reference sequence was derived from multiple replicate ESTs and validated by similar human genomic sequence. This gene encodes a member of a family of small membrane proteins that share a 35-amino acid signature sequence domain, beginning with the sequence PFXYD and containing 7 invariant and 6 highly conserved amino acids. The approved human gene nomenclature for the family is FXYD-domain containing ion transport regulator. Transmembrane topology has been established for two family members (FXYD1 and FXYD2), with the N-terminus extracellular and the C-terminus on the cytoplasmic side of the membrane. FXYD2, also known as the gamma subunit of the Na, K-ATPase, regulates the properties of that enzyme. FXYD1 (phospholemman), FXYD2 (gamma), FXYD3 (MAT-8), FXYD4 (CHIF), and FXYD5 (RIC) have been shown to induce channel activity in experimental expression systems. This gene product, FXYD7, is novel and has not been characterized as a protein.
Specifications
| FXYD7 | |
| Polyclonal | |
| Unconjugated | |
| FXYD7 | |
| 1110035I01Rik; FLJ25096; FXYD domain containing ion transport regulator 7; FXYD domain-containing ion transport regulator 7; FXYD7; Unknown (protein for MGC:128487) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 53822 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P58549 | |
| FXYD7 | |
| Synthetic peptide directed towards the N-terminal of human FXYD7 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction