missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FYB/ADAP/SLAP130 Antibody (460107) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB38571JF669
This item is not returnable.
View return policy
Description
FYB/ADAP/SLAP130 Monoclonal antibody specifically detects FYB/ADAP/SLAP130 in Human samples. It is validated for Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| FYB/ADAP/SLAP130 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| ADAP, Adhesion and degranulation promoting adaptor protein, adhesion and degranulation-promoting adaptor protein, FYB-120/130SLAP-130FYN-binding protein (FYB-120/130), FYN binding protein, FYN-binding protein, FYN-T-binding protein, p120/p130, PRO0823, SLAP130, SLP-76-associated phosphoprotein | |
| E. coli-derived recombinant human FYB/ADAP/SLAP130, Ala2-Gly274, Accession # O15117 | |
| 0.1 mL | |
| Immunology | |
| 2533 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry (Intracellular Staining), CyTOF | |
| 460107 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction