missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FYB/ADAP/SLAP130 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76810MFV450
This item is not returnable.
View return policy
Description
FYB/ADAP/SLAP130 Polyclonal antibody specifically detects FYB/ADAP/SLAP130 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| FYB/ADAP/SLAP130 | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| ADAP, Adhesion and degranulation promoting adaptor protein, adhesion and degranulation-promoting adaptor protein, FYB-120/130SLAP-130FYN-binding protein (FYB-120/130), FYN binding protein, FYN-binding protein, FYN-T-binding protein, p120/p130, PRO0823, SLAP130, SLP-76-associated phosphoprotein | |
| Antibody was raised against a 20 amino acid synthetic peptide from near the carboxy terminus of human ADAP. The immunogen is located within amino acids 640 - 690 of ADAP. Amino Acid Squence: LKGKDDRKKSIREKPKVSD | |
| 0.1 mL | |
| Immunology | |
| 2533 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction