missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FYCO1, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00079443-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: MNTKVTSDWYYARSPFLQPKLSSDIVGQLYELTEVQFDLASRGFDLDAAWPTFARRTLTTGSSAYLWKPPSRSSSMSSLVSSYLQTQEMVSNFDLNSPLNNEALEGFDEMRLELDQLEVREKQLQERMQQLDRENQELRAAVSQQGEQLQTERERGRTAAEDNVRLTCLVAELQKQWEVTQATQNTVKELQTCLQALELGAAEKEEDYHTALRRLESMLQPLAQELEATRDSLDKKNQHLASFPGWLAMALHVGDSpecifications
| FYCO1 | |
| Polyclonal | |
| 50 % glycerol | |
| BC007218 | |
| FYCO1 | |
| FYCO1 (AAH07218, 1 a.a. ∼ 255 a.a) full-length recombinant protein with GST tag. | |
| RUO | |
| 79443 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| FYVE and coiled-coil domain containing 1 | |
| DKFZp779K1152/FLJ13335/MGC126517/MGC126519/RUFY3/ZFYVE7 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction