missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FYCO1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104495
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FYCO1.
This gene encodes a protein that contains a RUN domain, FYVE-type zinc finger domain and Golgi dynamics (GOLD) domain. The encoded protein plays a role in microtubule plus end-directed transport of autophagic vesicles through interactions with the small GTPase Rab7, phosphatidylinositol-3-phosphate (PI3P) and the autophagosome marker LC3. Mutations in this gene are a cause of autosomal recessive congenital cataract-2 (CATC2).
Specifications
| FYCO1 | |
| Polyclonal | |
| Unconjugated | |
| FYCO1 | |
| 2810409M01Rik; CATC2; CTRCT18; DKFZp779K1152; FLJ13335; FYCO1; FYVE and coiled-coil domain autophagy adaptor 1; FYVE and coiled-coil domain containing 1; FYVE and coiled-coil domain-containing protein 1; maternal embryonic message 2; Mem2; MGC126517; MGC126519; RUFY3; RUN and FYVE domain containing 3; ZFYVE7; Zinc finger FYVE domain-containing protein 7 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 301085, 79443 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9BQS8 | |
| FYCO1 | |
| A synthesized peptide derived from human FYCO1(Accession Q9BQS8), corresponding to amino acid residues E832-C882. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction