missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fyn Polyclonal Antibody
GREENER_CHOICE

Product Code. 17722014
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17722014 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17722014 Supplier Invitrogen™ Supplier No. PA5120612

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse testis, Mouse brain, Rat brain. The target is usually found in the following locations: Cell membrane, Cytoplasm, Nucleus. Immunogen sequence: VKHYKIRKLD NGGYYITTRA QFETLQQLVQ HYSERAAGLC CRLVVPCHKG MPRLTDLSVK TKDVWEIPRE SLQLIKRLGN GQFGEVWMGT WNGNTKVAIK T.

FYN is a SRC-related kinase expressed in brain, skeletal muscle and adipose tissues. Fyn functions in a large number of pathways and has been shown to play a role in memory. FYN is a member of the protein-tyrosine kinase oncogene family. It encodes a membrane-associated tyrosine kinase that has been implicated in the control of cell growth. The protein associates with the p85 subunit of phosphatidylinositol 3-kinase and interacts with the fyn-binding protein. Alternatively spliced transcript variants encoding distinct isoforms exist.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fyn
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Fyn
Gene Accession No. P06241, P39688, Q62844
Gene Alias AI448320; AW552119; C syn protooncogene; c-syn protooncogene; EC 2.7.10.2; Fyn; fyn {ECO:0000312; FYN oncogene related to SRC, FGR, YES; Fyn proto-oncogene; FYN proto-oncogene, Src family tyrosine kinase; kinase Fyn; MGC45350; OKT3-induced calcium influx regulator; OTTHUMP00000017916; OTTHUMP00000017917; P59 p59-Fyn; p59-Fyn; Proto-oncogene c-Fyn; Protooncogene Syn; proto-oncogene Syn; proto-oncogene tyrosine-protein kinase Fyn; RGD:2641}; RP1-66H14.1; SLK; Src Kinase p59; Src like kinase; src/yes-related novel; src-like kinase; SYN; tyrosine kinase p59fyn(T); tyrosine-protein kinase Fyn
Gene Symbols Fyn
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 200-300 of human Fyn (NP_002028.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14360, 25150, 2534
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.