missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FYTTD1 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-04090DL594
This item is not returnable.
View return policy
Description
FYTTD1 Polyclonal antibody specifically detects FYTTD1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| FYTTD1 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| forty-two-three domain containing 1, Forty-two-three domain-containing protein 1, Protein 40-2-3, UAP56-interacting factor, UIFDKFZp761B1514 | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human UAP56-Interacting Factor using the numbering given in entry NP_115664.2 (GeneID 84248). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunoprecipitation | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 84248 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction