missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FYTTD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598705
This item is not returnable.
View return policy
Description
FYTTD1 Polyclonal Antibody for Western Blot, ELISA
Required for mRNA export from the nucleus to the cytoplasm. Acts as an adapter that uses the DDX39B/UAP56-NFX1 pathway to ensure efficient mRNA export and delivering to the nuclear pore. Associates with spliced and unspliced mRNAs simultaneously with ALYREF/THOC4. [UniProt].
Specifications
| FYTTD1 | |
| Polyclonal | |
| Unconjugated | |
| FYTTD1 | |
| 2010005M05Rik; 3830411L18Rik; 3830421F13Rik; 40-2-3 protein; 4933423A17Rik; Ac1176; AI098218; AW539567; C85190; forty-two-three domain containing 1; forty-two-three domain-containing protein 1; Forty-two-three domain-containing protein 1-like protein; FYTTD1; Protein 40-2-3; putative 40-2-3 protein; RGD1306899; UAP56-interacting factor; UAP56-interacting factor-like; UIF | |
| Rabbit | |
| Protein G | |
| RUO | |
| 84248 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q96QD9 | |
| FYTTD1 | |
| Recombinant Human UAP56-interacting factor protein (1-318AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction