missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FZD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581392
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Mouse FZD1/Frizzled-1.
Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD1 protein contains a signal peptide, a cysteine-rich domain in the N-terminal extracellular region, 7 transmembrane domains, and a C-terminal PDZ domain-binding motif. The FZD1 transcript is expressed in various tissues.
Specifications
| FZD1 | |
| Polyclonal | |
| Unconjugated | |
| Fzd1 | |
| AW227548; Chfz-1; Dfrizzled-1; DKFZp564G072; Drosophila polarity gene (frizzled) homologue; Drosophila polarity gene homolog 1; FLJ95923; frizzled; frizzled 1; frizzled 1, seven transmembrane spanning receptor; frizzled class receptor 1; frizzled family receptor 1; frizzled homolog 1; frizzled homolog 1 (Drosophila); frizzled, Drosophila, homolog of, 1; frizzled-1; Fz receptor 1; Fz1; FZ-1; Fzd1; FzE1; hFz1; mFz1; rFz1; Wnt receptor; Xfrizzled-1 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 14362 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with no preservative | |
| O70421 | |
| Fzd1 | |
| Recombinant Mouse FZD1 protein (Met1-His248). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction