missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FZD9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533564
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gibbon, Mouse, Rat, Hamster, Panda, Dog, Bovine, Bat, Rabbit, Opossum (100%) Platypus (86%).
Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD9 gene is located within the Williams syndrome common deletion region of chromosome 7, and heterozygous deletion of the FZD9 gene may contribute to the Williams syndrome phenotype. FZD9 is expressed predominantly in brain, testis, eye, skeletal muscle, and kidney.
Specifications
| FZD9 | |
| Polyclonal | |
| Unconjugated | |
| FZD9 | |
| CD349; CD349 antigen; Frizzled 3; frizzled 9; frizzled 9, seven transmembrane spanning receptor; frizzled class receptor 9; frizzled family receptor 9; frizzled homolog 9; frizzled homolog 9 (Drosophila); Frizzled3; Frizzled-3; Frizzled9; frizzled-9; Frizzled-like protein 9; fz-9; FZD3; FZD9; FzE6; hFz9; mFz3; mfz9; rFz9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100345803, 100761155, 14371, 266608, 489809, 521364, 8326 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| O00144, Q8K4C8, Q9R216 | |
| FZD9 | |
| Synthetic 14 amino acid peptide from 1st extracellular domain of human FZD9 / Frizzled 9. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit, Bovine, Hamster, Bat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction