missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FZD9 Recombinant Rabbit Monoclonal Antibody (23GB5475), Invitrogen™

Product Code. 30089164
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30089164 20 μL 20µL
30089756 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30089164 Supplier Invitrogen™ Supplier No. MA552863

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Members of the 'frizzled' gene family encode 7-transmembrane domain proteins that are receptors for Wnt signaling proteins. The FZD9 gene is located within the Williams syndrome common deletion region of chromosome 7, and heterozygous deletion of the FZD9 gene may contribute to the Williams syndrome phenotype. FZD9 is expressed predominantly in brain, testis, eye, skeletal muscle, and kidney.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FZD9
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5475
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FZD9
Gene Accession No. O00144
Gene Alias CD349; CD349 antigen; Frizzled 3; frizzled 9; frizzled 9, seven transmembrane spanning receptor; frizzled class receptor 9; frizzled family receptor 9; frizzled homolog 9; frizzled homolog 9 (Drosophila); Frizzled3; Frizzled-3; Frizzled9; frizzled-9; Frizzled-like protein 9; fz-9; FZD3; FZD9; FzE6; hFz9; mFz3; mfz9; rFz9
Gene Symbols FZD9
Host Species Rabbit
Immunogen A synthesized peptide derived from human Frizzled 9/CD349 (550-591AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8326
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.