Learn More
G-418 Disulfate, BLD Pharm™
Description
- Chemical Name: (2R,3S,4R,5R,6S)-5-Amino-6-(((1R,2S,3S,4R,6S)-4,6-diamino-3-(((2R,3R,4R,5R)-3,5-dihydroxy-5-methyl-4-(methylamino)tetrahydro-2H-pyran-2-yl)oxy)-2-hydroxycyclohexyl)oxy)-2-((R)-1-hydroxyethyl)tetrahydro-2H-pyran-3,4-diol bis(sulfate)
- Purity: ≥720u/mg
- CAS Number: 108321-42-2
- SMILES: O=S(O)(O)=O.O[C@@H]([C@@H]1O[C@@]([C@@H]([C@@H](O)[C@@H]2O)N)([H])O[C@]2([H])[C@H](O)C)[C@H]([C@@H](C[C@@H]1N)N)O[C@](OC[C@](C)(O)[C@@H]3NC)([H])[C@@H]3O.O=S(O)(O)=O
Specifications
Specifications
| CAS | 108321-42-2 |
| Molecular Formula | C20H44N4O18S2 |
| MDL Number | MFCD28168024 |
| Quantity | 1 g |
| SMILES | O=S(O)(O)=O.O[C@@H]([C@@H]1O[C@@]([C@@H]([C@@H](O)[C@@H]2O)N)([H])O[C@]2([H])[C@H](O)C)[C@H]([C@@H](C[C@@H]1N)N)O[C@](OC[C@](C)(O)[C@@H]3NC)([H])[C@@H]3O.O=S(O)(O)=O |
| Molecular Weight (g/mol) | 692.71 |
| Chemical Name or Material | G-418 disulfate |