missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G-CSF Antibody (7E4F7) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52447-0.025mg
This item is not returnable.
View return policy
Description
G-CSF Monoclonal specifically detects G-CSF in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| G-CSF | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000, CyTOF-ready | |
| C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3OS, filgrastim, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin | |
| Mouse | |
| 22.3 kDa | |
| 0.025 mg | |
| Biologically Active Proteins, Cell Cycle and Replication, Cytokine Research | |
| 1440 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2a |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| 7E4F7 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CSF3 | |
| Purified recombinant fragment of human CSF3 (AA: 1-207) expressed in E. Coli. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction