missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G-CSF Antibody (SPM468), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47933G
This item is not returnable.
View return policy
Description
G-CSF Monoclonal antibody specifically detects G-CSF in Human, Macaque samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Specifications
| G-CSF | |
| Monoclonal | |
| DyLight 488 | |
| C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3OS, filgrastim, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin | |
| Nuclei from pokeweed mitogen stimulated human peripheral blood lymphocytes | |
| 0.1 mL | |
| Cell Cycle and Replication, Cytokine Research | |
| 1440 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence | |
| SPM468 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Macaque | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction