missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G-CSFR/CD114 Antibody (38643), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3811E
This item is not returnable.
View return policy
Description
G-CSFR/CD114 Monoclonal antibody specifically detects G-CSFR/CD114 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA (Matched Antibody Pair)
Specifications
| G-CSFR/CD114 | |
| Monoclonal | |
| DyLight 405 | |
| CD114, CD114 antigen, colony stimulating factor 3 receptor (granulocyte), G-CSF receptor, GCSFRG-CSF-R, granulocyte colony-stimulating factor receptor | |
| Mouse myeloma cell line NS0-derived recombinant human G-CSF R/CD114, Glu25-Pro621, Accession # Q99062 | |
| 0.5 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 38643 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1441 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction