Learn More
G-CSFR/CD114 Antibody (723806), PerCP, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB60391C
Product Code. 18632924
  • ONLINE EXCLUSIVE PRICE
    € 670.00
    € 633.15 / 0.10mL
    Save € 36.85
    5% Off
Description
G-CSFR/CD114 Monoclonal antibody specifically detects G-CSFR/CD114 in Mouse samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
G-CSFR/CD114
Monoclonal
PerCP
PBS
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
1441
Store at 4°C in the dark.
IgG2a
Flow Cytometry, CyTOF
723806
Flow Cytometry, CyTOF-ready
CD114, CD114 antigen, colony stimulating factor 3 receptor (granulocyte), G-CSF receptor, GCSFRG-CSF-R, granulocyte colony-stimulating factor receptor
Mouse myeloma cell line NS0-derived recombinant mouse G-CSF R/CD114, Cys26-Asp626, Accession # P40223
0.1 mL