Learn More
IKA™ G-L PCR Tube Adapter
Product Code. 16278701 Shop All IKA Products
Change view
Click to view available options
For Use With (Equipment):
1.5 to 2 mL
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
16278701 1.5 to 2 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16278701 Supplier IKA™ Supplier No. 0030001346
Only null left
Add to basket
Add to basket
G-L PCR Tube Adapter for use with 1.5 to 2 mL rotor.
  • Adapter
  • PCR Tube
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 1.5 to 2 mL
Description Diameter: 6 mm
Includes 6/Pk