missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G-substrate Antibody (OTI2A8), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71645AF532
This item is not returnable.
View return policy
Description
G-substrate Monoclonal antibody specifically detects G-substrate in Human samples. It is validated for Western Blot
Specifications
| G-substrate | |
| Monoclonal | |
| Alexa Fluor 532 | |
| chromosome 7 open reading frame 16, GSBSG-substrate | |
| Full length recombinant protein of human G-substrate (NP_006649) produced in E.coli. | |
| 0.1 mL | |
| Signal Transduction | |
| 10842 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot | |
| OTI2A8 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction