missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G-substrate Antibody (OTI2A8) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71645
This item is not returnable.
View return policy
Description
G-substrate Monoclonal specifically detects G-substrate in Human samples. It is validated for Western Blot.
Specifications
| G-substrate | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000 | |
| chromosome 7 open reading frame 16, GSBSG-substrate | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 10842 | |
| Human | |
| Lyophilized |
| Western Blot | |
| OTI2A8 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| PPP1R17 | |
| Full length human recombinant protein of human C7ORF16 (NP_006649) produced in E.coli. | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction