missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G2A/GPR132 Rabbit anti-Human, Mouse, Monkey, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 186.00 - € 497.00
Spezifikation
| Antigen | G2A/GPR132 |
|---|---|
| Applications | Western Blot, ELISA, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
18112262
|
Novus Biologicals
NBP2-69090-0.025ML |
0.025 mL |
€ 186.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18614649
|
Novus Biologicals
NBP2-69090 |
100 μL |
€ 497.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
G2A/GPR132 Polyclonal specifically detects G2A/GPR132 in Human, Mouse, Monkey samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Spezifikation
| G2A/GPR132 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Monkey | |
| G protein-coupled receptor 132, G2 accumulation protein, G2AG protein-coupled receptor G2A, MGC99642, probable G-protein coupled receptor 132 | |
| Synthesized peptide derived from the C-terminal region of human G2A. | |
| Primary | |
| Store at -20°C. |
| Western Blot, ELISA, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| GPCR | |
| PBS containing 50% glycerol, 0.5% BSA | |
| 29933 | |
| IgG | |
| Affinity purified |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts