Learn More
G3BP1 Rabbit anti-Human, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL594-13057-100UL
Product Code. 17462633
  • € 450.00 / 100µL
Description
The G3BP1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human G3BP1. This antibody recognizes human, rat, mouse antigen. The G3BP1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
Specifications
G3BP1
Polyclonal
Rabbit
Primary
Human, Rat, Mouse
Immunofluorescence
CoraLite 594
100 μL
10146