missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G3BP1 Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 80777-1-RR-20UL
This item is not returnable.
View return policy
Description
The G3BP1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human G3BP1. This antibody recognizes human antigen. The G3BP1 antibody has been validated for the following applications: IHC, IF/ICC, IF-P, ELISA analysis.Specifications
| G3BP1 | |
| Recombinant Monoclonal | |
| 1000 μg/mL | |
| G3BP; HDH-VIII; MGC111040 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| Recombinant Antibody | |
| IgG |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunofluorescence, ELISA | |
| 7F14 | |
| Unconjugated | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 10146 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction